Skip to Content
Merck
All Photos(1)

Key Documents

Z409391

Sigma-Aldrich

Nalgene® safety waste systems

complete, capacity 10 L

Synonym(s):

laboratory waste liquid disposal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

manufacturer/tradename

Nalgene®

capacity

10 L

Looking for similar products? Visit Product Comparison Guide

General description

Safely dispose of chemical and biological waste and reduce hazardous emissions in the laboratory.

Funnels and waste bottles for the 4L model are also available, sold separately.

Safety waste funnel has a hinged cover (red) to keep emissions contained, funnel attaches to the bottle and remains in place until the bottle is full. A built-in vent minimizes overflow. Bottles are made of safe, durable HDPE.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service