Z407186
Aldrich® Sure/Seal™ septum-inlet transfer adapter
polypropylene, pkg of 1 ea
About This Item
Recommended Products
material
polypropylene
packaging
pkg of 1 ea
Related Categories
General description
The 8 mm septum cap (included) with flat PTFE-faced septum screws onto the side-arm for dispensing or short term storage. Standard rubber septa (Z565695 or Z565679) fit on side-arms for dispensing. For use with Aldrich products packed in Sure/Seal bottles.
Linkage
Customers Also Viewed
Articles
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.
Preserve reagent quality of air- and moisture-sensitive reagents using nitrogen or argon in crown-cap bottles with a 6 mm diameter hole in the crown-cap and a PTFE-faced rubber liner.