Z303941
Clips for Quickfit® flat-flange reaction flask lids
For 100 mm bore
Synonym(s):
JC100F
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
stainless steel
label
D
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Quickfit JC 100F
availability
not available in USA and Canada
General description
Designed for securing flat flange joints, these clips have a hinged ′spring wire′ design that tightens using a wing nut.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)