Key Documents
Z217085
Sigma-Aldrich
Scienceware® microtube cutter
for tubes up to 1.5 mL
About This Item
UNSPSC Code:
41121702
NACRES:
NB.11
manufacturer/tradename
Bel-Art F21018-0000
General description
Cutters are perfect for retrieving radioactive pellets from the tips of tubes without resuspension or the risk of splattered radioactive substance. Works best with polypropylene and polyethylene tubes up to 1.5ml, not recommended for use with polystyrene tubes. Maximum cut diameter: 9.4mm (.37"). Replacement blades available.
Safely and accurately removes microcentrifuge tube tips
  • Eliminating the hazards of using knives or razor blades
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)