WHA10318493
Whatman® qualitative filter paper, Grade 470
sheets, L × W 460 mm × 570 mm
Synonym(s):
Z749478
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
sheets
feature
Grade 470
packaging
pk of 100
manufacturer/tradename
Whatman 10318493
Whatman Article No. 28414407 (US reference)
parameter
fast flow rate
L × W
460 mm × 570 mm
pore size
20-30 μm
Looking for similar products? Visit Product Comparison Guide
General description
Grade 470 papers applications sheets.
Features and Benefits
- Soft surface
- For gelatinous samples.
- The absorption of culture media, as blotting paper, for electrophoresis and amino acid chromatography.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service