WHA10313032
Whatman® weighing boats and paper
609 Kjeldahl weighing boat, 100/pk
Synonym(s):
Z613592, weigh boat, weigh paper, weighing paper
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.71
Recommended Products
material
paper
feature
grade 609
grade B-2
packaging
pack of 100 ea
manufacturer/tradename
Cytiva
Whatman 10313032
Whatman Article No. 28414318 (US reference)
L × W
55 mm × 10 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Cytiva′s Whatman parchment weigh boats are designed so users can collect, transfer, and drop both sample and weighing boat into the acid solution during a Kjeldahl analysis without influencing analytical results.
Grade 609 weigh boats
Whatman weigh boats from Cytiva are approximately 0.07 mm thick. They come in packs of 100, with dimensions of 55 × 10 × 10 mm (2.165 × 0.39 × 0.39 inch).
Grade 609 weigh boats
Whatman weigh boats from Cytiva are approximately 0.07 mm thick. They come in packs of 100, with dimensions of 55 × 10 × 10 mm (2.165 × 0.39 × 0.39 inch).
Features and Benefits
- A Fast and reliable way to safely transfer samples for Kjeldahl analysis
- Excellent for both weighing and quantitative sample transfer
- Made from low nitrogen parchment paper
- No glue or chemical additives
- Whatman weigh boats dissolve residue-free in the digestion solution without influencing analytical results
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service