Skip to Content
Merck
All Photos(1)

Key Documents

W313602

Sigma-Aldrich

2,3-Diethylpyrazine

≥98%, FG

Synonym(s):

Shoyu pyrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19
FEMA Number:
3136
EC Number:
Council of Europe no.:
534
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.005
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

300
400

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 110

Assay

≥98%

refractive index

n20/D 1.5 (lit.)

bp

180-182 °C (lit.)

density

0.963 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

green; nutty; pepper

SMILES string

CCc1nccnc1CC

InChI

1S/C8H12N2/c1-3-7-8(4-2)10-6-5-9-7/h5-6H,3-4H2,1-2H3

InChI key

GZXXANJCCWGCSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

2,3-Diethylpyrazine has been identified as one of the volatile flavor constituents in roasted coffee beans, roasted sesame seeds and baked potato.

Application


  • Comparison of pyrazines formation in methionine/glucose and corresponding Amadori rearrangement product model.: This research explores the formation of pyrazines, including 2,3-diethylpyrazine, through the reactions of methionine and glucose, providing insights into the pathways and mechanisms involved in the generation of these compounds which are crucial for food flavoring (Deng et al., 2022).


Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chemistry of baked potato flavor. 1. Pyrazines and thiazoles indentified in the volatile flavor of baked potato.
Coleman EC & Ho CT.
Journal of Agricultural and Food Chemistry, 28(1), 66-68 (1980)
Analysis of roasted coffee bean volatiles by using comprehensive two-dimensional gas chromatography?time-of-flight mass spectrometry
Ryan D, et al.
Journal of Chromatography A, 1054(1), 57-65 (2004)
Effect of antioxidants on the volatiles of roasted sesame seeds.
Soliman MA, et al.
Journal of Agricultural and Food Chemistry, 33(3), 523-528 (1985)

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service