Skip to Content
Merck
All Photos(3)

Key Documents

W244317

Sigma-Aldrich

Ethyl 2-methylbutyrate

greener alternative

natural, mixture of enantiomers, ≥98%, FCC, FG

Synonym(s):

Berry butyrate, Ethyl 2-methylbutanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)COOC2H5
CAS Number:
Molecular Weight:
130.18
FEMA Number:
2443
Beilstein:
1720887
EC Number:
Council of Europe no.:
265c
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.409
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
food allergen:
no known allergens

grade

FG
Fragrance grade
Halal
Kosher
natural

Quality Level

300
400

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117

Assay

≥98%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.397 (lit.)

bp

133 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

apple; green; fruity

SMILES string

CCOC(=O)C(C)CC

InChI

1S/C7H14O2/c1-4-6(3)7(8)9-5-2/h6H,4-5H2,1-3H3

InChI key

HCRBXQFHJMCTLF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 2-methylbutyrate is a volatile flavor compound found in stawberries, Scheurebe and Gewu1 rztraminer wines.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Flavor quality of cultivated strawberries: the role of the sulfur compounds
Dirinck PJ, et al.
Journal of Agricultural and Food Chemistry, 29(2), 316-321 (1981)
Identification of character impact odorants of different white wine varieties.
Guth H.
Journal of Agricultural and Food Chemistry, 45(8), 3022-3026 (1997)
Johannes Niebler et al.
Phytochemistry, 109, 66-75 (2014-12-04)
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile
Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not
Helene Hopfer et al.
Molecules (Basel, Switzerland), 20(5), 8453-8483 (2015-05-20)
Twenty-seven commercial Californian Cabernet Sauvignon wines of different quality categories were analyzed with sensory and chemical methods. Correlations between five quality proxies-points awarded during a wine competition, wine expert scores, retail price, vintage, and wine region-were correlated to sensory attributes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service