Skip to Content
Merck
All Photos(1)

Key Documents

W220906

Sigma-Aldrich

Butyl phenylacetate

≥98%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CO2(CH2)3CH3
CAS Number:
Molecular Weight:
192.25
FEMA Number:
2209
EC Number:
Council of Europe no.:
2159
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.787
NACRES:
NA.21
Organoleptic:
honey; rose
grade:
FG
Fragrance grade
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 172.515

Assay

≥98%

refractive index

n20/D 1.49 (lit.)

bp

133-135 °C/15 mmHg (lit.)

density

0.99 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

honey; rose

SMILES string

CCCCOC(=O)Cc1ccccc1

InChI

1S/C12H16O2/c1-2-3-9-14-12(13)10-11-7-5-4-6-8-11/h4-8H,2-3,9-10H2,1H3

InChI key

LDOXTQYWWYXYSQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service