TEM-CF200F1-AU-UL
Ultral-Thin Carbon Film Supported Gold Gilder Finder F1 Grids
size 200 mesh, box of 50
All Photos(1)
About This Item
Recommended Products
packaging
box of 50
size
200 mesh
General description
A 3-4 nm ultral-thin film of pure carbon supported glider finder (F1) grids. Gold grid, 200 mesh. Gilder finder grids are locator grids with reference/index on the grid: 3.05mm diameter, 0.7 mil (18μm) thickness. F1 type of design has thick bars dividing regions into 6 thin bar areas, which are identified by a numeric system.
Choose from one of the most recent versions:
Certificates of Analysis (COA)