Skip to Content
Merck
All Photos(1)

Documents

T5605

Sigma-Aldrich

α,α,α′,α′-Tetrabromo-o-xylene

97%

Synonym(s):

1,2-Bis(dibromomethyl)benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CHBr2)2
CAS Number:
Molecular Weight:
421.75
Beilstein:
2211698
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

114-116 °C (lit.)

SMILES string

BrC(Br)c1ccccc1C(Br)Br

InChI

1S/C8H6Br4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4,7-8H

InChI key

LNAOKZKISWEZNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α,α,α′,α′-Tetrabromo-o-xylene can be employed as a precursor for the synthesis of:
  • Poly(o-phenylene vinylene) (o-PPV) via electrochemical polymerization.
  • A pentacene derivative by reacting with naphthalene moieties for the development of organic thin-film transistors (OTFTs).
  • Bitopic pyrazole containing ligands.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new type of soluble pentacene precursor for organic thin-film transistors
Chen K-Y, et al.
Chemical Communications (Cambridge, England), 118(10), 1065-1067 (2007)
On the electrochemical polymerization of poly (p-phenylene vinylene) and poly (o-phenylene vinylene)
Peres LO, et al.
Synthetic Metals, 118(1-3), 65-70 (2001)
Synthesis of new bitopic tetra (pyrazolyl)-ligands with neopentane and o-xylene backbones
Potapov AS, et al.
TheScientificWorldJournal, 2012(10), 1065-1067 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service