SYNA792434
Synthware™ bushing adapter
female joint: ST/NS 24/40, male joint: ST/NS 34/45
Synonym(s):
Synthware A792434, glass bushing adapter
All Photos(1)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.51
Recommended Products
Joint
female joint: ST/NS24/40
male joint: ST/NS34/45
packaging
pkg of 1 ea
manufacturer/tradename
Synthware A792434
General description
Bushing adapters allow for compact assemblies of dissimilar joints.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)