Key Documents
MT51107686
Mettler Toledo DX series ISE half-cell membrane kit
For Cadmium ISE
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
manufacturer/tradename
Mettler-Toledo 51107686
General description
DX series ion specific membrane kit. Mount the new membrane onto the shaft of your ISE half-cell, and you have a new ISE!
Kit contains one membrane plus electrolyte
Choose from one of the most recent versions:
Certificates of Analysis (COA)