Skip to Content
Merck
All Photos(1)

Key Documents

DWKCP0010P

Azlon Hexagonal Base Polypropylene Cylinders with Molded Graduations

class B, capacity 10 mL , with +/-0.2mL tolerance

Synonym(s):

CP0010P

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121805
NACRES:
NA.54

feature

class B
graduated

packaging

pkg of 10 ea

manufacturer/tradename

Azlon CP0010P

capacity

10 mL , with +/-0.2mL tolerance

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the measurement and dispensing of liquids in the laboratory

Features and Benefits

Conform to ISO 6706 with anti-suction designto prevent base sticking to wet surface

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service