DWKBWH0060PN
Azlon HDPE Wide Neck Round Bottles
screw cap, capacity 60 mL
Synonym(s):
BWH0060PN
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
Recommended Products
material
screw cap
wide-neck
packaging
pkg of 10 ea
manufacturer/tradename
Azlon BWH0060PN
capacity
60 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for a wide range of liquid storage and sampling applications
Features and Benefits
Suitable for food contact applications
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service