Key Documents
CLS72502L
Pyrex® Mallory direct reading settlometer
volume 2000 mL
Synonym(s):
lab equipment accessory
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
manufacturer/tradename
Corning 7250-2L
volume
2000 mL
Related Categories
General description
Used with oxidized sludge process for treatment of sewage and other wastes. 2000 mL heavy wall cylinder measures 51/8". I.D. × 71/2". high and is graduated in cc per liter, and in hundredths of a foot.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)