CLS65640200
PYREXPLUS® volumetric flask, with protective coating, Pyrex® stopper
capacity 200 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass, Corning® 65640
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
long
stopper size
13
sterility
non-sterile
feature
closure type ST stopper
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 65640-200
tol. measuring range
0.10 mL
capacity
200 mL
General description
Flask features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121 °C) and resistant to thermal shock.
Caution
Do not place over direct heat or flame. Do not heat above 121 °C moist heat or 110 °C dry heat.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)