CLS5564010
Pyrex® class A low-actinic volumetric flask, Pyrex® ST stopper
capacity 10 mL
Synonym(s):
flasks glass, volumetric flasks, volumetric glass
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
borosilicate glass
long
Joint
joint stopper9
sterility
non-sterile
feature
closure type ST stopper
packaging
case of 12
manufacturer/tradename
Corning 55640-10
Corning 55640
tol. measuring range
0.02 mL
capacity
10 mL
General description
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of flask, which retains mechanical strength, chemical stability, and thermal resistance of Pyrex brand labware.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)