Key Documents
CLS3953312C
Pyrex® gas-dispersion tube
pore size 40-60 μm
Synonym(s):
Corning® 39533
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
manufacturer/tradename
Corning 39533-12C
pore size
40-60 μm
General description
With 12 mm O.D. fritted cylinder, 250 mm L × 8 mm diam. stem. Used to disperse gas in liquids. Size of bubbles depends on porosity. Size permits insertion through small openings.
Features and Benefits
  • Pyrex® gas-dispersion tube is made of Type I Class A - Low-Expansion Borosilicate Glass.
  • Coarse with 40-60 μm pore size.
  • May withstand autoclaving.
  • Not designed for vacuum and freezing applications.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)