Skip to Content
Merck
All Photos(1)

Key Documents

CLS139525

Pyrex® round media storage bottles and reusable screw caps

capacity 25 mL

Synonym(s):

Pyrex® media bottles, glass media bottles, laboratory bottles, round bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

borosilicate glass bottle
clear bottle
cylindrical bottle
orange polypropylene cap (plug seal)
wide-mouth bottle

sterility

non-sterile

feature

autoclavable
cap
graduations
reusable

packaging

case of 10

manufacturer/tradename

Corning 1395-25

availability

not available in EU

capacity

25 mL

diam. × H

36.5 mm × 70 mm

fitting

GL 25 neck thread

Looking for similar products? Visit Product Comparison Guide

General description

These heavy duty 25 mL PYREX round bottles can be used for storage as well as mixing and sampling. They include a linerless, one-piece autoclavable orange polypropylene plug seal GL25 threaded cap. A pouring ring is not included on the 25 mL size. Optional red PBT caps and red ETFE pouring rings are available for dry heat sterilization (180°C). Bottles have permanent white enamel graduations and marking spots.

Caution: DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.
Pyrex heavy-duty round media storage bottles can be used for storage as well as for mixing and sampling. These media storage bottles Include linerless, one-piece, autoclavable, orange PP plug-seal caps with drip-free pouring rings. Optional red PBTP caps and red ETFE pouring rings are available separately, for dry heat sterilization (180°C).

Pyrex round media storage bottles are made of borosilicate glass with permanent, white enamel graduations and marking spots.

Caution: Bottles larger than 2 L should NOT be used with bottle-top filter units or in other applications involving vacuum/pressure, as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage.

A wide range of optional caps are available for these bottles:
  • Vented membrane cap with 0.22 μm PTFE hydrophobic membrane
  • Red PBT (polybutylene terephthalate) solid caps and ETFE pouring rings for dry heat sterilization (180°C)
  • PBT caps with silicone septa for syringe sampling or introduction of reagents

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service