CLS1000PACK
PYREX® Griffin beakers
low form, assorted pack (contains 1 beaker of 5 different sizes)
Synonym(s):
Pyrex® beakers, Pyrex® glass beakers, beakers, glass beakers, Corning® 1000
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass
feature
graduations
packaging
case of 1
manufacturer/tradename
Corning 1000-PACK
availability
not available in EU
General description
PYREX® assortment pack of Griffin Low form beakers, double scale, graduated A convenience pack of PYREX beakers contains one of each of the most popular sizes. Designed for the low volume user, a case contains one of each beaker: 50mL, 100mL, 250mL, 600mL and 1L. Packaged in a partitioned carton for safe transit and storage.
Low Form (Corning 1000 series) Griffin beakers have the following features:
- Graduated for approximate content
- Double-graduated metric scale
- Beakers have a spout
- Extra-large area for marking
- Uniform wall thickness
- Heavy duty, industrial grade glass beakers
Other Notes
Browse all glass beakers
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.