All Photos(3)
About This Item
Linear Formula:
C4H6(=O)
CAS Number:
Molecular Weight:
70.09
Beilstein:
1560289
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.421 (lit.)
bp
99 °C (lit.)
density
0.938 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
O=C1CCC1
InChI
1S/C4H6O/c5-4-2-1-3-4/h1-3H2
InChI key
SHQSVMDWKBRBGB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Starting material for a new, efficient synthesis of aminocyclobutanecarboxylic acid via the intermediacy of sulfonyloxiranes.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
60.8 °F
Flash Point(C)
16 °C
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tetrahedron, 62, 9268-9268 (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service