Skip to Content
Merck
All Photos(2)

Key Documents

C60600

Sigma-Aldrich

4-Chloro-3-nitrobenzotrifluoride

97%

Synonym(s):

2-Chloro-5-(trifluoromethyl)nitrobenzene, 4-Chloro-3-nitro-α,α,α-trifluorotoluene, CMNT, MNT-Cl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)CF3
CAS Number:
Molecular Weight:
225.55
Beilstein:
523986
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.489 (lit.)

bp

222 °C (lit.)

density

1.511 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1cc(ccc1Cl)C(F)(F)F

InChI

1S/C7H3ClF3NO2/c8-5-2-1-4(7(9,10)11)3-6(5)12(13)14/h1-3H

InChI key

TZGFQIXRVUHDLE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Subchronic oral toxicity in the rat of 3-nitro-4-chloro-alpha,alpha,alpha-trifluorotoluene (NCTT)].
R Bucchi et al.
Annali dell'Istituto superiore di sanita, 19(2-3), 385-389 (1983-01-01)
[Acute oral toxicity in the rat of 3-nitro-4-chloro-alpha,alpha, alpha-trifluorotoluene (NCTT) and 3,5-dinitro-4-chloro-alpha,alpha, alpha-trifluorotoluene (DNCTT)].
A R Bucchi et al.
Annali dell'Istituto superiore di sanita, 19(2-3), 351-354 (1983-01-01)
Joshua Kurek et al.
Environmental science & technology, 53(13), 7316-7325 (2019-06-13)
Historic forest management practices led to widespread aerial application of insecticides, such as dichlorodiphenyltrichloroethane (DDT), to North American conifer forests during ∼1950-1970. Lake basins thus may provide an important archive of inputs and aquatic responses to these organochlorines. We use
In vitro mutational studies with trifluralin and trifluorotoluene derivatives.
R Benigni et al.
Annali dell'Istituto superiore di sanita, 18(1), 123-126 (1982-01-01)
S Giorgini et al.
Italian general review of dermatology, 15(2), 107-112 (1978-05-01)
A case of occupational contact dermatitis in a female chemical pharmaceutical laboratory researcher is described. The allergological investigation revealed a state of sensitization to three mono-nitrobenzenes with halogen substitutions in the nucleus or a side chain. Problems related to sensitization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service