Documents
BR781510
Brand®1.2 ML Tubes
with 12 strips of 8 tubes, polypropylene
Synonym(s):
Sample Storage, Tube racks, Tubes
About This Item
UNSPSC Code:
41122804
NACRES:
NB.22
material
polypropylene
sterility
non-sterile
feature
with 12 strips of 8 tubes
manufacturer/tradename
BRAND 781510
packaging
pkg of (10 racks with 12 strips of 8 tubes)
capacity
1.2 mL
General description
PP. Tubes and rack are autoclavable at 121 °C (2 bar), acc. DIN EN 285. Ideal for biological tests, such as PCR, cell uptake studies, RIA and EIA. Designed for storing and freezing down to -80 °C, and for transporting reagents and samples. Compatible with standard microplates and suitable for working with multichannel pipettes. (Size of tubes: outer Ø 8.8 mm, height 45 mm.) Single caps and strip caps, PE, available as accessories.BIO-CERT® CERTIFIED QUALITY: Free of human DNA, RNase, DNase and Pyrogens
Features and Benefits
  • BRAND® 1.2 ML tubes racks are manufactured by high-quality polypropylene.
  • Designed for storing and transporting reagents.
  • Suitable for biological tests, such as PCR, cell growth studies, RIA, and EIA.
  • Tubes and racks are autoclavable at 121°C.
  • Temperature resistant down to -80°C.
  • Tight sealing closure options prevents sample loss.
  • Compact design is ideal for use in automated or multi-channel systems.
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG