Key Documents
BR718905
BRAND® counting chamber BLAUBRAND® Bürker pattern
without clips, double ruled
Synonym(s):
BLAUBRAND®, Bürker, Cell culture, Counting Chambers
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
Pricing and availability is not currently available.
material
colorless
glass
sterility
non-sterile
feature
without clips
manufacturer/tradename
BRAND 718905
wells
1
General description
Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.The ruling shows 9 large squares of 1 mm² each. These are used for counting leucocytes.Each large square is subdivided by double lines (0.05 mm apart) into 16 group squares with 0.2 mm sides. The group squares correspond in size to the Neubauer counting chamber, but have no further subdivisions. They are used for counting thrombocytes and erythrocytes. The double lines from mini squares with an area of 0.0025 mm².
Features and Benefits
• BLAUBRAND® quality• Outstanding measurement precision• 100% certified quality
Packaging
1 counting chamber, complete with 2 haemacytometer cover glasses, in transparent plastic box.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)