Skip to Content
Merck
All Photos(1)

Key Documents

BR137755

BRAND® magnetic stirring bar retriever, PP

diam. 10 mm, L 450 mm, with ring

Synonym(s):

Magnetic stirring bar retriever, Magnetic stirring bars

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122402
NACRES:
NB.72

feature

with ring

manufacturer/tradename

BRAND 137755

L

450 mm

diam.

10 mm

General description

The BRAND® Magnetic Stirring Bar Retriever is an essential tool for efficiently retrieving magnetic stirring bars from containers and vessels. Constructed from durable polypropylene (PP), this retriever features a fully sealed magnetic core.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service