BMSBR5000
Everlast™ rocker with flat mat
AC/DC input 115 V AC, US 3-pin plug
All Photos(1)
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
manufacturer/tradename
Benchmark Scientific BR5000
W × D
16 in. × 12 in.
Related Categories
General description
- Stainless-steel platform
- Dual platform accessory doubles work surface
- Cold room and incubator safe
The rocking speed and tilt angle can easily be adjusted
to vary between gentle and aggressive agitation. Both
the standard platform and the (optional) stacking
platform are constructed of stainless steel, preventing
against any rusting or corrosion. The Everlast 247 is
designed to provide years of uninterrupted service.
Legal Information
Everlast is a trademark of Benchmark Scientific, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)