Skip to Content
Merck
All Photos(1)

Documents

A94654

Sigma-Aldrich

12-Aminododecanolactam

98%

Synonym(s):

Aza-2-cyclotridecanone, Cyclododecanone isooxime, Laurolactam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23NO
CAS Number:
Molecular Weight:
197.32
Beilstein:
122031
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

150-153 °C (lit.)

SMILES string

O=C1CCCCCCCCCCCN1

InChI

1S/C12H23NO/c14-12-10-8-6-4-2-1-3-5-7-9-11-13-12/h1-11H2,(H,13,14)

InChI key

JHWNWJKBPDFINM-UHFFFAOYSA-N

General description

12-aminododecanolactam is a cyclic amide that is used in a variety of organic synthesis reactions. It is a versatile building block that can be used to synthesize a wide range of compounds, including polyamides, peptides and amino acids.

Application

12-Aminododecanolactam can be used to synthesis low viscosity lactam-based ionic liquids by reacting with various Bronsted acids. It is also used to synthesis Nylon-12, by ring opening polymerization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

383.0 °F - closed cup

Flash Point(C)

195 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 12-aminododecenoic acid by coupling transaminase to oxylipin pathway enzymes
Coenen, et al.
Applied Microbiology and Biotechnology, 107, 2209-2221 (2023)
Low viscosity lactam-based ionic liquids with carboxylate anions: Synthesis, characterization, thermophysical properties and mutual miscibility of ionic liquid with alcohol, water, and hydrocarbons
Bessa, et al.
Journal of Molecular Liquids, 313, 113586-113586 (2020)
Magdalena Kwiatkowska et al.
Polymers, 12(2) (2020-02-08)
In this work the preparation of polyamide 12 (PA12) based composites reinforced with pristine and surface-modified carbon nanotubes is reported. A qualitative and quantitative evaluation of multi-walled carbon nanotube functionalization with oxygen containing reactive groups achieved by different procedures of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service