Skip to Content
Merck
All Photos(2)

Key Documents

A4380

Sigma-Aldrich

Ammonium biborate tetrahydrate

98.9-101.2% B2O3 basis

Synonym(s):

Ammonium tetraborate tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2B4O7 · 4H2O
CAS Number:
Molecular Weight:
263.38
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Assay

98.9-101.2% B2O3 basis

form

powder

pH

~8.8 ( in solution, over a wide range of concentrations)

SMILES string

N.N.O.O.O.O.OB1OB(O1)OB2OB(O)O2

InChI

1S/B4H2O7.2H3N.4H2O/c5-1-7-3(8-1)11-4-9-2(6)10-4;;;;;;/h5-6H;2*1H3;4*1H2

InChI key

QZNWNKFWDJRMLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Determination of nucleation kinetics of ammonium biborate tetrahydrate: This study focuses on the metastable zone widths (MZW) of ammonium biborate tetrahydrate, both in its pure state and in the presence of impurities, providing insights into its crystallization kinetics and stability (O Sahin et al., 2014).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service