Skip to Content
Merck
All Photos(1)

Key Documents

930881

Sigma-Aldrich

(cod)Pd(CH2CMe2C6H4)

≥95%

Synonym(s):

Campora’s palladacycle

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24Pd
CAS Number:
Molecular Weight:
346.80
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.21

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: catalyst
reaction type: Cross Couplings

Application

(cod)Pd(CH2CMe2C6H4), also referred to as Campora′s palladacycle, is a Pd(II) source with demonstrated use as an air-, moisture-, and thermally stable precursor to palladium oxidative addition complexes (Pd OACs).

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan P King et al.
Organic letters, 23(20), 7927-7932 (2021-10-07)
The utilization of isolated Palladium Oxidative Addition Complexes (OACs) has had a significant impact on Pd-catalyzed and Pd-mediated cross-coupling reactions. Despite their importance, widespread utility of OACs has been limited by the instability of their precursor complexes. Herein, we report

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service