912182
Enantioprobe (S,S)-7
≥95%
Synonym(s):
N-((1S,2S)-2-(Benzyloxy)cyclopentyl)-3-(3-(but-3-yn-1-yl)-3H-diazirin-3-yl)propanamide, Fully functionalized enantioprobe, Ligandability probe
About This Item
Recommended Products
Quality Level
Assay
≥95%
form
powder
storage temp.
−20°C
SMILES string
N([C@@H]2[C@H](CCC2)OCc3ccccc3)C(=O)CCC1(N=N1)CCC#C
InChI
1S/C20H25N3O2/c1-2-3-13-20(22-23-20)14-12-19(24)21-17-10-7-11-18(17)25-15-16-8-5-4-6-9-16/h1,4-6,8-9,17-18H,3,7,10-15H2,(H,21,24)/t17-,18-/m0/s1
InChI key
JWDFLBJRNHEKSI-ROUUACIJSA-N
Application
Together, the 16 enantioprobes support ligandability studies in living cells, a significant method for development of chemical probes and lead discovery efforts to find binders for traditionally ″″undruggable″″ protein targets. This strategy is also compatible with multiplexing for higher throughput.
- Fluorophore-conjugated azide tags for SDS-PAGE in-gel profiling: 910147, 760757, 760765, 777315, 777323
- Azide-biotin tags for enrichment: 901765, 914134, 914460, 762024, 900912, 900891, QBD10825
- Azide capture solid supports: 900957, 742627
- Streptavidin for biotin enrichment: S6940, E5529, E2513, 69203, 16-126, S1638
- Mass spectrometry and Multiplexing
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service