Skip to Content
Merck
All Photos(1)

Key Documents

908568

Sigma-Aldrich

[Ir(dFCF3ppy)2-(5,5′-dCF3bpy)]PF6

≥95%

Synonym(s):

[5,5′-Bis(trifluoromethyl)-2,2′-bipyridine-N1,N1′]bis[3,5-difluoro-2-[5-(trifluoromethyl)-2-pyridinyl-N]phenyl-C]Iridium(III) hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H16F22IrN4P
CAS Number:
Molecular Weight:
1145.69
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

mp

>300 °C

photocatalyst activation

460 nm

SMILES string

[Ir+](c5c(c(cc(c5)F)F)c6ncc(cc6)C(F)(F)F)c3c(c(cc(c3)F)F)c4ncc(cc4)C(F)(F)F.FC(F)(F)c1cnc(cc1)c2ncc(cc2)C(F)(F)F.F[P-](F)(F)(F)(F)F

InChI

1S/C12H6F6N2.2C12H5F5N.F6P.Ir/c13-11(14,15)7-1-3-9(19-5-7)10-4-2-8(6-20-10)12(16,17)18;2*13-8-2-3-9(10(14)5-8)11-4-1-7(6-18-11)12(15,16)17;1-7(2,3,4,5)6;/h1-6H;2*1-2,4-6H;;/q;;;-1;+1

InChI key

QEXYETWHTSSWGM-UHFFFAOYSA-N

Application

[Ir(dFCF3ppy)2-(5,5′-dCF3bpy)]PF6 is a cyclometalated iridium(III) complex that can be used in visible-light mediated photocatalytic organic transformations including the alkylation of remote C-H bonds and alkene aminoarylation.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy M Monos et al.
Science (New York, N.Y.), 361(6409), 1369-1373 (2018-09-29)
Alkene aminoarylation with a single, bifunctional reagent is a concise synthetic strategy. We report a catalytic protocol for the addition of arylsulfonylacetamides across electron-rich alkenes with complete anti-Markovnikov regioselectivity and excellent diastereoselectivity to provide 2,2-diarylethylamines. In this process, single-electron alkene
Gilbert J Choi et al.
Nature, 539(7628), 268-271 (2016-11-04)
Despite advances in hydrogen atom transfer (HAT) catalysis, there are currently no molecular HAT catalysts that are capable of homolysing the strong nitrogen-hydrogen (N-H) bonds of N-alkyl amides. The motivation to develop amide homolysis protocols stems from the utility of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service