Skip to Content
Merck
All Photos(1)

Key Documents

857416

Sigma-Aldrich

(−)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate

99%, for peptide synthesis

Synonym(s):

α-Methyl-L-DOPA, L-α-Methyl-DOPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2C(CH3)(NH2)CO2H · 11/2H2O
CAS Number:
Molecular Weight:
238.24
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Product Name

(−)-3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate, 99%

Quality Level

Assay

99%

form

powder

optical activity

[α]25/D −13°, c = 1 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

O.O.O.C[C@](N)(Cc1ccc(O)c(O)c1)C(O)=O.C[C@](N)(Cc2ccc(O)c(O)c2)C(O)=O

InChI

1S/2C10H13NO4.3H2O/c2*1-10(11,9(14)15)5-6-2-3-7(12)8(13)4-6;;;/h2*2-4,12-13H,5,11H2,1H3,(H,14,15);3*1H2/t2*10-;;;/m00.../s1

InChI key

YKFCISHFRZHKHY-NGQGLHOPSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Ostrov et al.
The Journal of clinical investigation, 128(5), 1888-1902 (2018-02-14)
Major histocompatibility (MHC) class II molecules are strongly associated with many autoimmune disorders. In type 1 diabetes (T1D), the DQ8 molecule is common, confers significant disease risk, and is involved in disease pathogenesis. We hypothesized that blocking DQ8 antigen presentation
Sebastiaan P van Kessel et al.
Nature communications, 10(1), 310-310 (2019-01-20)
Human gut microbiota senses its environment and responds by releasing metabolites, some of which are key regulators of human health and disease. In this study, we characterize gut-associated bacteria in their ability to decarboxylate levodopa to dopamine via tyrosine decarboxylases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service