767700
Mytilus edulis foot protein-1
1 mg/mL (in 1% citric acid), sterile
Synonym(s):
MAPcell
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
liquid
concentration
1 mg/mL (in 1% citric acid)
refractive index
n20/D 1.335 (lit.)
bp
100-103 °C/760 mmHg
density
1.008 g/mL at 25 °C (lit.)
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
A solution of MEFP-1 that is specially developed for biological applications. The protein can be used to coat surfaces in order to facilitate immobilization of cells and tissue. The solution is sterile by autoclaving and has an endotoxin level of = 1 EU/ml. The product is cell culture tested (cytotoxicity and cell adhesion assay).
MEFP-1 can be successfully used for unspecific immobilization of cells and tissue to essentially any surface. It is an especially useful tool when the surface chemistry results in poor adhesion of cells or for cells that are difficult to attach. In comparison to binding strategies based on electrostatic interactions (positively charged surfaces) or covalent immobilization (EDC/NHS or glutaraldehyde) attachment using MEFP-1 is insensitive to ionic strength and pH and will not result in undesired chemical modification, such as cross-linking, of cell surface components. The protein coating is a useful tool in techniques such as in situ hybridization, immunohistochemistry, tissue engineering, drug delivery, ELISA, microfluidics, microinjection and various microscopy techniques.
MEFP-1 can be successfully used for unspecific immobilization of cells and tissue to essentially any surface. It is an especially useful tool when the surface chemistry results in poor adhesion of cells or for cells that are difficult to attach. In comparison to binding strategies based on electrostatic interactions (positively charged surfaces) or covalent immobilization (EDC/NHS or glutaraldehyde) attachment using MEFP-1 is insensitive to ionic strength and pH and will not result in undesired chemical modification, such as cross-linking, of cell surface components. The protein coating is a useful tool in techniques such as in situ hybridization, immunohistochemistry, tissue engineering, drug delivery, ELISA, microfluidics, microinjection and various microscopy techniques.
Suitability
MAPcell is sterile and suitable for cell culture.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
No data available
Flash Point(C)
No data available
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
J H Waite et al.
Science (New York, N.Y.), 212(4498), 1038-1040 (1981-05-29)
The fouling marine mussel Mytilus edulis attaches itself to various substrates by spinning byssal threads, the adhesive discs of which are rich in the amino acid 3,4-dihydroxyphenylalanine (dopa). An acid-soluble protein was extracted and purified from the phenol gland located
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service