Skip to Content
Merck
All Photos(1)

Key Documents

764086

Sigma-Aldrich

5-Bromo-2,1,3-benzothiadiazole

95%

Synonym(s):

5-Bromobenzo[c][1,2,5]thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3BrN2S
CAS Number:
Molecular Weight:
215.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

solid

mp

55-60 °C

storage temp.

2-8°C

SMILES string

Brc1ccc2nsnc2c1

InChI

1S/C6H3BrN2S/c7-4-1-2-5-6(3-4)9-10-8-5/h1-3H

InChI key

LLCRUZDFDGTAAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service