726303
N1,N4-Bis-Boc-spermidine
≥95.0% (TLC)
Synonym(s):
1,5-Di-Boc-1,5,10-triazadecane, N1,N4Di-Boc-1,8-diamino-4-azaoctane, {3-[(4-Aminobutyl)-tert-butoxycarbonylamino]propyl}carbamic acid tert-butyl ester
About This Item
Recommended Products
Assay
≥95.0% (TLC)
form
liquid
reaction suitability
reagent type: cross-linking reagent
storage temp.
−20°C
SMILES string
CC(C)(C)OC(=O)NCCCN(CCCCN)C(=O)OC(C)(C)C
InChI
1S/C17H35N3O4/c1-16(2,3)23-14(21)19-11-9-13-20(12-8-7-10-18)15(22)24-17(4,5)6/h7-13,18H2,1-6H3,(H,19,21)
InChI key
GQZZMJDEWJRWRP-UHFFFAOYSA-N
Related Categories
Packaging
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service