710490
Bis(isopropylcyclopentadienyl)manganese
97%
Synonym(s):
1,1′-Bis(1-methylethyl) manganocene
About This Item
Recommended Products
Assay
97%
form
liquid
reaction suitability
core: manganese
reagent type: catalyst
density
1.090 g/cm3 at 25 °C
SMILES string
[Mn].CC(C)[C]1[CH][CH][CH][CH]1.CC(C)[C]2[CH][CH][CH][CH]2
InChI
1S/2C8H11.Mn/c2*1-7(2)8-5-3-4-6-8;/h2*3-7H,1-2H3;
InChI key
CRDYYRDPRXAYHD-UHFFFAOYSA-N
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Guidelines on use of proteinase K, an enzyme commonly used to degrade proteins, and protect DNA and RNA from degradation in samples.
Guidelines on use of proteinase K, an enzyme commonly used to degrade proteins, and protect DNA and RNA from degradation in samples.
Guidelines on use of proteinase K, an enzyme commonly used to degrade proteins, and protect DNA and RNA from degradation in samples.
Balancing Proteinase K cost with quality and technical support ensures optimal enzyme selection for diverse applications.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service