Skip to Content
Merck
All Photos(2)

Key Documents

706213

Sigma-Aldrich

1-(2-Morpholinoethyl)-1H-pyrazole-4-boronic acid pinacol ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26BN3O3
CAS Number:
Molecular Weight:
307.20
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

powder

mp

89-94 °C

functional group

ether

SMILES string

CC1(C)OB(OC1(C)C)c2cnn(CCN3CCOCC3)c2

InChI

1S/C15H26BN3O3/c1-14(2)15(3,4)22-16(21-14)13-11-17-19(12-13)6-5-18-7-9-20-10-8-18/h11-12H,5-10H2,1-4H3

InChI key

FBAPTUAEBQMVEY-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service