Skip to Content
Merck
All Photos(1)

Key Documents

69493

Sigma-Aldrich

Methyl triphenylmethyl ether

≥97.0% (HPLC)

Synonym(s):

Methyl trityl ether, Trityl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3COCH3
CAS Number:
Molecular Weight:
274.36
Beilstein:
1914067
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

crystals

mp

81-83 °C

functional group

ether

SMILES string

COC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C20H18O/c1-21-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19/h2-16H,1H3

InChI key

IRMNIXXVOOMKKP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service