Skip to Content
Merck
All Photos(1)

Key Documents

689734

Sigma-Aldrich

2-Chloro-4-pyridinecarboxaldehyde

technical, ≥90%

Synonym(s):

2-Chloro-4-formylpyridine, 2-Chloroisonicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4ClNO
CAS Number:
Molecular Weight:
141.56
Beilstein:
7986625
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Quality Level

Assay

≥90%
≥90.0% (GC)

form

lumps

functional group

aldehyde
chloro

SMILES string

Clc1cc(C=O)ccn1

InChI

1S/C6H4ClNO/c7-6-3-5(4-9)1-2-8-6/h1-4H

InChI key

UFPOSTQMFOYHJI-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service