Skip to Content
Merck
All Photos(3)

Key Documents

687650

Sigma-Aldrich

But-3-enylboronic acid

≥95%

Synonym(s):

3-Butenylboronic acid, But-1-ene-4-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2CH2B(OH)2
CAS Number:
Molecular Weight:
99.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

form

powder

mp

84-90 °C

functional group

allyl

storage temp.

2-8°C

SMILES string

OB(O)CCC=C

InChI

1S/C4H9BO2/c1-2-3-4-5(6)7/h2,6-7H,1,3-4H2

InChI key

WHTULVNQGYJCSG-UHFFFAOYSA-N

General description

May contain varying amounts of anhydride.

Application

Reactant for:
  • Ag(I)-promoted Suzuki-Miyaura cross-couplings

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service