Skip to Content
Merck
All Photos(1)

Key Documents

674575

Sigma-Aldrich

MacMillan Imidazolidinone OrganoCatalysts Kit I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352005
eCl@ss:
32161000
NACRES:
NA.22

General description

MacMillan imidazolidinone organocatalysts are metal-free chiral catalysts widely used in enantioselective organic transformations. These organocatalysts are non-toxic, low cost, and robust in simple reaction conditions because of their insensitivity to air and moisture.

Application

MacMillan Imidazolidinone organocatalysts can be used to catalyze various organocatalytic transformation reactions such as 1,3-dipolar cycloadditions, Diels–Alder reaction, Friedel-Crafts alkylations, α-chlorinations, α-fluorinations, and intramolecular Michael reactions to access a high level of enantioselective products. It can also be used in enantioselective photocatalytic reactions, hydride reduction, and Mukaiyama-Michael reaction.

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 569763(5S)-(−)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone monohydrochloride, 97% 500 mgSDS

  • 661902(S)-(+)-2-(tert-Butyl)-3-methyl-4-imidazolidinone trifluoroacetic acid, 96% 500 mgSDS

  • 663085(5S)-(−)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone dichloroacetic acid, 97% 500 mgSDS

  • 663107(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone, 97% 500 mgSDS

  • 668540(2S,5S)-(–)-5-Benzyl-3-methyl-2-(5-methyl-2-furyl)-4-imidazolidinone, 95% 250 mgSDS

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent Development of Recoverable MacMillan Catalyst in Asymmetric Organic Transformations
Singh S
Advanced Synthesis & Catalysis, 363(3), 629-656 (2021)
Imidazolidinones as Asymmetric Organocatalysts
Sustainable Catalysis: Without Metals or Other Endangered Elements (2015)
Recent Development of Recoverable MacMillan Catalyst in Asymmetric Organic Transformations
Singh S
advanced synthesis and catalysis, 363(3), 629-656 (2021)

Articles

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service