659657
Poly(ethylene glycol)-block-polylactide methyl ether
PEG average Mn 750, PLA average Mn 1,000
Synonym(s):
PEG-b-PLA
About This Item
Recommended Products
mol wt
PEG average Mn 750
PLA average Mn 1,000
Quality Level
Looking for similar products? Visit Product Comparison Guide
Features and Benefits
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service