Skip to Content
Merck
All Photos(1)

Documents

600032

Sigma-Aldrich

Allyl alcohol-2-13C

99 atom % 13C

Synonym(s):

2-Propen-2-13C-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=13CHCH2OH
CAS Number:
Molecular Weight:
59.07
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

99% (CP)

bp

96-98 °C (lit.)

mp

-129 °C (lit.)

density

0.869 g/mL at 25 °C

mass shift

M+1

SMILES string

OC[13CH]=C

InChI

1S/C3H6O/c1-2-3-4/h2,4H,1,3H2/i2+1

InChI key

XXROGKLTLUQVRX-VQEHIDDOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing Lyu et al.
Proceedings of the National Academy of Sciences of the United States of America, 116(2), 546-555 (2018-12-26)
SENCR is a human-specific, vascular cell-enriched long-noncoding RNA (lncRNA) that regulates vascular smooth muscle cell and endothelial cell (EC) phenotypes. The underlying mechanisms of action of SENCR in these and other cell types is unknown. Here, levels of SENCR RNA
Lena Reimann et al.
Communications biology, 3(1), 253-253 (2020-05-24)
The PI3K/Akt pathway promotes skeletal muscle growth and myogenic differentiation. Although its importance in skeletal muscle biology is well documented, many of its substrates remain to be identified. We here studied PI3K/Akt signaling in contracting skeletal muscle cells by quantitative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service