538078
Propionyl chloride-functionalized silica gel
200-400 mesh, extent of labeling: 1 mmol/g loading
Synonym(s):
Carboxychloro-2-functionalized silica gel
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
extent of labeling
1 mmol/g loading
surface area
500 m2/g
particle size
200-400 mesh
pore size
60 Å pore size
storage temp.
2-8°C
SMILES string
O=[SiH2].CCC(Cl)=O
InChI
1S/C3H5ClO.H2OSi/c1-2-3(4)5;1-2/h2H2,1H3;2H2
InChI key
IYPMNWVACSXYDT-UHFFFAOYSA-N
Application
Scavenges primary & secondary amines and alcohols
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service