Skip to Content
Merck
All Photos(1)

Key Documents

524964

Sigma-Aldrich

N-Methylfurfurylamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.471 (lit.)

bp

148-152 °C (lit.)

density

0.982 g/mL at 25 °C (lit.)

SMILES string

CNCc1ccco1

InChI

1S/C6H9NO/c1-7-5-6-3-2-4-8-6/h2-4,7H,5H2,1H3

InChI key

DGLIOWSKNOCHEX-UHFFFAOYSA-N

General description

N-Methylfurfurylamine is a secondary amine. It can be prepared by reacting furfural and aqueous trimethylamine.

Application

N-Methylfurfurylamine may be used in the synthesis of cis -α-[2-[(2-furanylmet hyl)methylamino]cyclohexyl]-benzenemethanol.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

105.1 °F - closed cup

Flash Point(C)

40.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of 6-substituted purines.
Bullock MW, et al.
Journal of the American Chemical Society, 78(15), 3693-3696 (1956)
An unusual isomerization of a furan-containing compound.
Szmuszkovicz J, et al.
The Journal of Organic Chemistry, 51(25), 5002-5005 (1986)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service