488542
Chloromethane-13C
99 atom % 13C
Synonym(s):
Methyl-13C chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
13CH3Cl
CAS Number:
Molecular Weight:
51.48
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.12
Recommended Products
vapor density
1.74 (vs air)
vapor pressure
3796 mmHg ( 20 °C)
isotopic purity
99 atom % 13C
Assay
99% (CP)
bp
−24.2 °C (lit.)
mp
−97 °C (lit.)
density
0.933 g/mL at 25 °C (lit.)
mass shift
M+1
SMILES string
[13CH3]Cl
InChI
1S/CH3Cl/c1-2/h1H3/i1+1
InChI key
NEHMKBQYUWJMIP-OUBTZVSYSA-N
Related Categories
Packaging
1 L quantity is packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressure at 21ºC is 19 psig for 1 L . This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Recommended products
Stainless steel control valves Z146951 or Z187224
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Carc. 2 - Flam. Gas 1 - Press. Gas Liquefied gas - STOT RE 2 Inhalation
Storage Class Code
2A - Gases
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service