Skip to Content
Merck
All Photos(1)

Documents

451916

Sigma-Aldrich

Sulfur-34S

90 atom % 34S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
34S
CAS Number:
Molecular Weight:
33.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

isotopic purity

90 atom % 34S

Quality Level

mp

119 °C (lit.)

application(s)

environmental

mass shift

M+2

SMILES string

[34S]

InChI

1S/S/i1+2

InChI key

NINIDFKCEFEMDL-NJFSPNSNSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service