445231
2-Methoxyphenylboronic acid
95%
Synonym(s):
(2-Methoxyphenyl)boric acid, 2-Methoxybenzeneboronic acid, o-Anisylboronic acid, o-Methoxybenzeneboronic acid, o-Methoxyphenylboronic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3OC6H4B(OH)2
CAS Number:
Molecular Weight:
151.96
Beilstein:
3030981
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.06
Recommended Products
Quality Level
Assay
95%
form
solid
mp
105-110 °C (lit.)
SMILES string
COc1ccccc1B(O)O
InChI
1S/C7H9BO3/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5,9-10H,1H3
InChI key
ROEQGIFOWRQYHD-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ji Gwang Yu et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(63), 16044-16050 (2017-08-24)
Four dibenzofuran-type host materials substituted with a carbazolylcarbazole moiety were synthesized to investigate the effect of substitution position on the material parameters and device performances of host materials. The carbazolylcarbazole moiety was substituted at the 1-, 2-, 3-, and 4-positions
Steffen Bugge et al.
European journal of medicinal chemistry, 94, 175-194 (2015-03-15)
An approach for optimization of epidermal growth factor receptor tyrosine kinase (EGFR-TK) inhibitors using truncated thienopyrimidine structures combined with enzymatic assay has been evaluated. This was done by synthesis and EGFR activity measurement of a series of fragment structures and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service