Skip to Content
Merck
All Photos(3)

Documents

438308

Sigma-Aldrich

Tetrabutylammonium sulfate solution

50 wt. % in H2O

Synonym(s):

N,N,N-tributyl-1-butanaminium sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[[CH3(CH2)3]4N]2SO4
CAS Number:
Molecular Weight:
580.99
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

50 wt. % in H2O

refractive index

n20/D 1.417

bp

84 °C

density

1.01 g/mL at 25 °C

SMILES string

[O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/2C16H36N.H2O4S/c2*1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h2*5-16H2,1-4H3;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

ZXUCBXRTRRIBSO-UHFFFAOYSA-L

Application


  • Dimethyl sulfate and diisopropyl sulfate as practical and versatile O-sulfation reagents: This article discusses the comparison of tetrabutylammonium sulfate with other sulfation reagents, underlining its versatility and efficiency in organic synthesis, especially in sulfation reactions (Yue et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sara J Lupton et al.
Journal of agricultural and food chemistry, 63(51), 10988-10994 (2015-12-20)
Perfluorooctane sulfonate (PFOS) is used in consumer products as a surfactant and is found in industrial and consumer waste, which ends up in wastewater treatment plants (WWTPs). PFOS does not breakdown during WWTP processes and accumulates in the biosolids. Common
Minmeng Zhao et al.
Journal of agricultural and food chemistry, 66(29), 7684-7691 (2018-07-06)
This study aims to investigate in ovo feeding (IOF) of creatine pyruvate (CrPyr) on glucose metabolism, hormone concentration, and the 5'-AMP-activated protein kinase (AMPK) pathway in breast muscle of embryos and neonatal broilers. The three treatments were noninjected control, 0.75%
Nesta Bortey-Sam et al.
Analytica chimica acta, 1098, 125-132 (2020-01-18)
Cyanide (both HCN and CN- are represented by CN) has multiple industrial applications, is commonly found in some foods, and is a component of fire smoke. Upon exposure, CN blocks production of adenosine triphosphate, causing cellular hypoxia and cytotoxic anoxia
Paul P Van Veldhoven et al.
Frontiers in cell and developmental biology, 8, 144-144 (2020-04-09)
Mice lacking PMP34, a peroxisomal membrane transporter encoded by Slc25a17, did not manifest any obvious phenotype on a Swiss Webster genetic background, even with various treatments designed to unmask impaired peroxisomal functioning. Peroxisomal α- and β-oxidation rates in PMP34 deficient
Sophie Geboers et al.
International journal of pharmaceutics, 485(1-2), 131-137 (2015-03-10)
Tenofovir-disoproxil-fumarate (TDF) is a double ester prodrug which enables intestinal uptake of tenofovir (TFV) after oral administration in humans. In this study, prodrug stability was monitored in situ in the human intestine and in vitro using biorelevant media. In fasted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service